missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethyl-1,3-phenylenediamine 98.0+%, TCI America™

Catalog No. p-7136691
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T127525G 25 g
T1275100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T127525G Supplier TCI America Supplier No. T127525G

Chemical Identifiers

CAS 3102-70-3
Molecular Formula C9H14N2
Molecular Weight (g/mol) 150.23
MDL Number MFCD00010149
InChI Key ZVDSMYGTJDFNHN-UHFFFAOYSA-N
Synonym 2,4-Diaminomesitylene, 2,4-Diamino-1,3,5-trimethylbenzene
PubChem CID 76547
IUPAC Name 2,4,6-trimethylbenzene-1,3-diamine
SMILES CC1=CC(C)=C(N)C(C)=C1N

Specifications

Melting Point 91°C
Quantity 25 g
Formula Weight 150.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trimethyl-1,3-phenylenediamine
EINECSNumber (3)-3812
RTECSNumber CZ1607000
TSCA No