missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxybenzeneboronic acid, 98%

Catalog No. AAL1983703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1983703 1 g
AAL1983706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1983703 Supplier Thermo Scientific Chemicals Supplier No. L1983703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 135159-25-0 | C9H13BO5 | 212.01 g/mol

2,4,6-Trimethoxybenzeneboronic acid is used as an intermediate in organic syntheses and in Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethoxybenzeneboronic acid is used as an intermediate in organic syntheses and in Suzuki reaction.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 135159-25-0
Molecular Formula C9H13BO5
Molecular Weight (g/mol) 212.01
MDL Number MFCD01114642
InChI Key PKLRXZVPEQJTTJ-UHFFFAOYSA-N
Synonym 2,4,6-trimethoxybenzeneboronic acid, 2,4,6-trimethoxyphenyl boronic acid, boronic acid,b-2,4,6-trimethoxyphenyl, 1,3,5-trimethoxybenzene-2-boronic ester, pubchem9566, 1,3,5-trimethoxybenzene-2-bronic ester, acmc-1cirz, ablock ab-12-9177, 2,4,6-trimethoxybenzeneboronicacid
PubChem CID 4197996
IUPAC Name (2,4,6-trimethoxyphenyl)boronic acid
SMILES COC1=CC(OC)=C(B(O)O)C(OC)=C1

Specifications

Melting Point 96°C to 100°C
Quantity 1 g
Beilstein 4315581
Solubility Information Slightly soluble in water.
Formula Weight 212.01
Percent Purity 98%
Chemical Name or Material 2,4,6-Trimethoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only