missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxybenzaldehyde 98.0+%, TCI America™

Catalog No. T26515G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26515G 5 g
T265125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26515G Supplier TCI America Supplier No. T26515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 830-79-5
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00003313
InChI Key CRBZVDLXAIFERF-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4,6-trimethoxy, 2,4,6-trimethoxybenz aldehyde, pubchem8264, acmc-1cuhn, ksc490m2h, 2,4,6-trimethoxy-benzaldehyde, 2,4,6-trimethoxybenz-aldehyde, 2,4,6-trimethoxy benz aldehyde, crbzvdlxaiferf-uhfffaoysa, 1-formyl-2,4,6-trimethoxybenzene
PubChem CID 70019
IUPAC Name 2,4,6-trimethoxybenzaldehyde
SMILES COC1=CC(=C(C(=C1)OC)C=O)OC

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 5 g
Formula Weight 196.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trimethoxybenzaldehyde
RTECSNumber CU8460200
TSCA No