missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxybenzaldehyde, 98%

Catalog No. AAA1123818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1123818 50 g
AAA1123809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1123818 Supplier Thermo Scientific Chemicals Supplier No. A1123818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 830-79-5 | C10H12O4 | 196.202 g/mol

2,4,6-Trimethoxybenzaldehyde was used in the preparation and characterization of three RNA-specific fluorescent probes and their use in live cell imaging. It was used as starting reagent for the regioselective synthesis of new (±)-8-alkyl-5,7-dihydroxy-4-(4-hydroxyphenyl)-3,4-dihydrocoumarins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethoxybenzaldehyde was used in the preparation and characterization of three RNA-specific fluorescent probes and their use in live cell imaging. It was used as starting reagent for the regioselective synthesis of new (+/-)-8-alkyl-5,7-dihydroxy-4-(4-hydroxyphenyl)-3,4-dihydrocoumarins.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 830-79-5
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00003313
InChI Key CRBZVDLXAIFERF-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4,6-trimethoxy, 2,4,6-trimethoxybenz aldehyde, pubchem8264, acmc-1cuhn, ksc490m2h, 2,4,6-trimethoxy-benzaldehyde, 2,4,6-trimethoxybenz-aldehyde, 2,4,6-trimethoxy benz aldehyde, crbzvdlxaiferf-uhfffaoysa, 1-formyl-2,4,6-trimethoxybenzene
PubChem CID 70019
IUPAC Name 2,4,6-trimethoxybenzaldehyde
SMILES COC1=CC(=C(C(=C1)OC)C=O)OC

Specifications

Melting Point 119°C to 123°C
Quantity 50 g
Beilstein 1956051
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material 2,4,6-Trimethoxybenzaldehyde
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 212-598-2
RTECSNumber CU8460200
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only