missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. T26545G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26545G 5 g
T26541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26545G Supplier TCI America Supplier No. T26545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 154549-38-9
Molecular Formula C15H25BO2
Molecular Weight (g/mol) 248.173
MDL Number MFCD02683099
InChI Key FGFYEKLWZBTLEW-UHFFFAOYSA-N
Synonym 2,4,6-triisopropylphenyl boronic acid, 2,4,6-triisopropylphenylboronic acid, 2,4,6-triisopropylbenzeneboronic acid, 2,4,6-tri propan-2-yl phenyl boronic acid, 2,4,6-tris propan-2-yl phenyl boronic acid, acmc-209dav, 2,4,6-triisopropylphenyiboronic acid, 2,4,6-triisopropylphenyl boronicacid
PubChem CID 15153544
IUPAC Name [2,4,6-tri(propan-2-yl)phenyl]boronic acid
SMILES B(C1=C(C=C(C=C1C(C)C)C(C)C)C(C)C)(O)O

Specifications

Melting Point 165°C
Color White
Quantity 5 g
Formula Weight 248.17
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Triisopropylphenylboronic Acid (contains varying amounts of Anhydride)