missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzoyl chloride, 98+%

Catalog No. AAA1128014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1128014 25 g
AAA1128006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1128014 Supplier Thermo Scientific Chemicals Supplier No. A1128014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 57199-00-5 | C16H23ClO | 266.809 g/mol

2,4,6-Triisopropylbenzoyl chloride is used as pharmaceuticcal intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Triisopropylbenzoyl chloride is used as pharmaceuticcal intermediates.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Incompatible with strong oxidizing agents, ammonia etc,.

Chemical Identifiers

CAS 57199-00-5
Molecular Formula C16H23ClO
Molecular Weight (g/mol) 266.809
MDL Number MFCD00015030
InChI Key OSKNTKJPGKHDHV-UHFFFAOYSA-N
Synonym 2,4,6-triisopropylbenzoyl chloride, benzoyl chloride, 2,4,6-tris 1-methylethyl, acmc-20aoev, 2,4,6-triiso-propylbenzoyl chloride, 2,4,6-tri propan-2-yl benzoyl chloride, benzoyl chloride,2,4,6-tris 1-methylethyl, 2,4,6-tris 1-methylethyl benzoic acid chloride
PubChem CID 92697
IUPAC Name 2,4,6-tri(propan-2-yl)benzoyl chloride
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)C(=O)Cl)C(C)C

Specifications

Melting Point 78°C to 82°C
Quantity 25 g
UN Number UN3261
Beilstein 2269534
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 266.81
Percent Purity ≥98%
Chemical Name or Material 2,4,6-Triisopropylbenzoyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 260-614-1
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only