CAS RN 57199-00-5
IUPAC Name:
2,4,6-tris(propan-2-yl)benzoyl chloride
Synonyms:
2,4,6-triisopropylbenzoyl chloride
Molecular Weight (g/mol):
266.81
Molecular Formula:
C16H23ClO
InChi Key:
OSKNTKJPGKHDHV-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC(C(C)C)=C(C(Cl)=O)C(=C1)C(C)C
1
–
1
of
1
results
1
Promotions Available
2,4,6-Triisopropylbenzoyl chloride, 98+%
2,4,6-Triisopropylbenzoyl chloride, 98+%
CAS: 57199-00-5 Molecular Formula: C16H23ClO Molecular Weight (g/mol): 266.809 MDL Number: MFCD00015030 InChI Key: OSKNTKJPGKHDHV-UHFFFAOYSA-N Synonym: 2,4,6-triisopropylbenzoyl chloride,benzoyl chloride, 2,4,6-tris 1-methylethyl,acmc-20aoev,2,4,6-triiso-propylbenzoyl chloride,2,4,6-tri propan-2-yl benzoyl chloride,benzoyl chloride,2,4,6-tris 1-methylethyl,2,4,6-tris 1-methylethyl benzoic acid chloride PubChem CID: 92697 IUPAC Name: 2,4,6-tri(propan-2-yl)benzoyl chloride SMILES: CC(C)C1=CC(=C(C(=C1)C(C)C)C(=O)Cl)C(C)C