missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Triisopropylbenzenesulfonyl Chloride 97.0+%, TCI America™
Supplier: TCI America T0459500G
Specifications
| 2,4,6-Triisopropylbenzenesulfonyl Chloride | |
| 96°C | |
| C15H23ClO2S | |
| 500 g | |
| 2,4,6-triisopropylbenzenesulfonyl chloride, 2,4,6-triisopropylbenzene-1-sulfonyl chloride, trisyl chloride, 2,4,6-triisopropylbenzenesulphonyl chloride, benzenesulfonyl chloride, 2,4,6-tris 1-methylethyl, 2,4,6-triisopropylbenzenesulfonylchloride, 2,4,6-triisopropyl-benzenesulfonyl chloride, tpscl 2,4,6-triisopropyl benzenesulfonyl chloride, 2,4,6-triisopropylphenylsulfonyl chloride, 2,4,6-triisopropyl benzenesulfonyl chloride | |
| CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)Cl)C(C)C | |
| 302.857 | |
| 302.86 | |
| Crystalline Powder |
| 6553-96-4 | |
| White | |
| MFCD00007433 | |
| 3261 | |
| JAPYIBBSTJFDAK-UHFFFAOYSA-N | |
| 2,4,6-tri(propan-2-yl)benzenesulfonyl chloride | |
| 81042 | |
| ≥97.0% (T) |
Chemical Identifiers
| 6553-96-4 | |
| 302.857 | |
| JAPYIBBSTJFDAK-UHFFFAOYSA-N | |
| 81042 | |
| CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)Cl)C(C)C |
| C15H23ClO2S | |
| MFCD00007433 | |
| 2,4,6-triisopropylbenzenesulfonyl chloride, 2,4,6-triisopropylbenzene-1-sulfonyl chloride, trisyl chloride, 2,4,6-triisopropylbenzenesulphonyl chloride, benzenesulfonyl chloride, 2,4,6-tris 1-methylethyl, 2,4,6-triisopropylbenzenesulfonylchloride, 2,4,6-triisopropyl-benzenesulfonyl chloride, tpscl 2,4,6-triisopropyl benzenesulfonyl chloride, 2,4,6-triisopropylphenylsulfonyl chloride, 2,4,6-triisopropyl benzenesulfonyl chloride | |
| 2,4,6-tri(propan-2-yl)benzenesulfonyl chloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator