missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzenesulfonyl Chloride 97.0+%, TCI America™

Catalog No. T045925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T045925G 25 g
T0459500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T045925G Supplier TCI America Supplier No. T045925G

Chemical Identifiers

CAS 6553-96-4
Molecular Formula C15H23ClO2S
Molecular Weight (g/mol) 302.857
MDL Number MFCD00007433
InChI Key JAPYIBBSTJFDAK-UHFFFAOYSA-N
Synonym 2,4,6-triisopropylbenzenesulfonyl chloride, 2,4,6-triisopropylbenzene-1-sulfonyl chloride, trisyl chloride, 2,4,6-triisopropylbenzenesulphonyl chloride, benzenesulfonyl chloride, 2,4,6-tris 1-methylethyl, 2,4,6-triisopropylbenzenesulfonylchloride, 2,4,6-triisopropyl-benzenesulfonyl chloride, tpscl 2,4,6-triisopropyl benzenesulfonyl chloride, 2,4,6-triisopropylphenylsulfonyl chloride, 2,4,6-triisopropyl benzenesulfonyl chloride
PubChem CID 81042
IUPAC Name 2,4,6-tri(propan-2-yl)benzenesulfonyl chloride
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)Cl)C(C)C

Specifications

Melting Point 96°C
Color White
Quantity 25 g
UN Number 3261
Formula Weight 302.86
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Triisopropylbenzenesulfonyl Chloride
TSCA No
Recommended Storage Refrigerator