missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzenesulfonyl chloride, 97%

Catalog No. p-205691
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC152790250 25 g Glass bottle
AC152791000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC152790250 Supplier Thermo Scientific Chemicals Supplier No. 152790250
Only null left

CAS: 6553-96-4 | C15H23ClO2S | 302.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6553-96-4
Molecular Formula C15H23ClO2S
Molecular Weight (g/mol) 302.85
MDL Number MFCD00007433
InChI Key JAPYIBBSTJFDAK-UHFFFAOYSA-N
Synonym 2,4,6-triisopropylbenzenesulfonyl chloride, 2,4,6-triisopropylbenzene-1-sulfonyl chloride, trisyl chloride, 2,4,6-triisopropylbenzenesulphonyl chloride, benzenesulfonyl chloride, 2,4,6-tris 1-methylethyl, 2,4,6-triisopropylbenzenesulfonylchloride, 2,4,6-triisopropyl-benzenesulfonyl chloride, tpscl 2,4,6-triisopropyl benzenesulfonyl chloride, 2,4,6-triisopropylphenylsulfonyl chloride, 2,4,6-triisopropyl benzenesulfonyl chloride
PubChem CID 81042
IUPAC Name 2,4,6-tri(propan-2-yl)benzenesulfonyl chloride
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)Cl)C(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 93°C to 97°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula [(CH3)2CH]3C6H2SO2Cl
Quantity 25 g
Fieser 01,1228; 03,308; 06,622
Solubility Information Solubility in water: decomposes
Formula Weight 302.85
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 2, 4, 6-Triisopropylbenzenesulfonyl chloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 229-479-6
TSCA TSCA

RUO – Research Use Only