missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzenesulfonyl Azide (wetted with ca. 10% Water) (unit weight on dry weight basis) 98.0+%, TCI America™

Catalog No. T34341G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34341G 1 g
T34345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34341G Supplier TCI America Supplier No. T34341G
Only null left

Chemical Identifiers

CAS 36982-84-0
Molecular Formula C15H23N3O2S
Molecular Weight (g/mol) 309.428
MDL Number MFCD00859286
InChI Key AEMWUHCKKDPRSK-UHFFFAOYSA-N
Synonym 2,4,6-triisopropylbenzenesulfonyl azide, 2,4,6-triisopropylbenzene-sulfonyl azide, trisyl azide, 2,4,6-triisopropylbenzene-sulfonyl azide,, benzenesulfonyl azide, 2,4,6-tris 1-methylethyl, 2,4,6-triisopropyl-benzenesulfonyl azide, n-diazo-2,4,6-triisopropyl-benzenesulfonamide, 2,4,6-tris 1-methylethyl-benzenesulfonyl azide, 2,4,6-triisopropylbenzenesulfonyl azide, stab. with ca water, trisyl azide solution
PubChem CID 371707
IUPAC Name N-diazo-2,4,6-tri(propan-2-yl)benzenesulfonamide
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)N=[N+]=[N-])C(C)C

Specifications

Melting Point 43°C
Color White-Yellow
Quantity 1 g
Formula Weight 309.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Triisopropylbenzenesulfonyl Azide (wetted with ca. 10% Water) (unit weight on dry weight basis
TSCA No
Recommended Storage Refrigerator