missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzeneboronic acid, 98%

Catalog No. AAB2289103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2289103 1 g
AAB2289106 5 g
AAB2289114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2289103 Supplier Thermo Scientific Chemicals Supplier No. B2289103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 154549-38-9 | C15H25BO2 | 248.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 154549-38-9
Molecular Formula C15H25BO2
Molecular Weight (g/mol) 248.173
MDL Number MFCD02683099
InChI Key FGFYEKLWZBTLEW-UHFFFAOYSA-N
Synonym 2,4,6-triisopropylphenyl boronic acid, 2,4,6-triisopropylphenylboronic acid, 2,4,6-triisopropylbenzeneboronic acid, 2,4,6-tri propan-2-yl phenyl boronic acid, 2,4,6-tris propan-2-yl phenyl boronic acid, acmc-209dav, 2,4,6-triisopropylphenyiboronic acid, 2,4,6-triisopropylphenyl boronicacid
PubChem CID 15153544
IUPAC Name [2,4,6-tri(propan-2-yl)phenyl]boronic acid
SMILES B(C1=C(C=C(C=C1C(C)C)C(C)C)C(C)C)(O)O

Specifications

Melting Point ∼165°C (decomposition)
Quantity 1 g
Formula Weight 248.16
Percent Purity 98%
Chemical Name or Material 2,4,6-Triisopropylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only