missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Triiodophenol 98.0+%, TCI America™
$177.11 - $537.13
Chemical Identifiers
| CAS | 609-23-4 |
|---|---|
| Molecular Formula | C6H3I3O |
| Molecular Weight (g/mol) | 471.802 |
| MDL Number | MFCD00002179 |
| InChI Key | VAPDZNUFNKUROY-UHFFFAOYSA-N |
| Synonym | phenol, 2,4,6-triiodo, triiodophenol, unii-9rb2r81a7u, 2,4,6-trijodfenol, 2,4,6-trijodfenol czech, 2,4,6-triiodo-phenol, chembl81721, 2,4,6-triiodo-phenol tip, 2,6-triiodophenol, phenol,4,6-triiodo |
| PubChem CID | 11862 |
| IUPAC Name | 2,4,6-triiodophenol |
| SMILES | C1=C(C=C(C(=C1I)O)I)I |
Chemical Identifiers
| 609-23-4 | |
| 471.802 | |
| VAPDZNUFNKUROY-UHFFFAOYSA-N | |
| 11862 | |
| C1=C(C=C(C(=C1I)O)I)I |
| C6H3I3O | |
| MFCD00002179 | |
| phenol, 2,4,6-triiodo, triiodophenol, unii-9rb2r81a7u, 2,4,6-trijodfenol, 2,4,6-trijodfenol czech, 2,4,6-triiodo-phenol, chembl81721, 2,4,6-triiodo-phenol tip, 2,6-triiodophenol, phenol,4,6-triiodo | |
| 2,4,6-triiodophenol |
Specifications
| 609-23-4 | |
| White | |
| MFCD00002179 | |
| phenol, 2,4,6-triiodo, triiodophenol, unii-9rb2r81a7u, 2,4,6-trijodfenol, 2,4,6-trijodfenol czech, 2,4,6-triiodo-phenol, chembl81721, 2,4,6-triiodo-phenol tip, 2,6-triiodophenol, phenol,4,6-triiodo | |
| C1=C(C=C(C(=C1I)O)I)I | |
| 471.802 | |
| 471.80 | |
| Crystalline Powder |
| 160°C | |
| C6H3I3O | |
| 5 g | |
| VAPDZNUFNKUROY-UHFFFAOYSA-N | |
| 2,4,6-triiodophenol | |
| 11862 | |
| ≥98.0% (HPLC,T) | |
| 2,4,6-Triiodophenol |
Safety and Handling
RTECSNumber : SN2800000
TSCA : Yes