missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triiodophenol 98.0+%, TCI America™

Catalog No. T04525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T04525G 5 g
T045225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T04525G Supplier TCI America Supplier No. T04525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 609-23-4
Molecular Formula C6H3I3O
Molecular Weight (g/mol) 471.802
MDL Number MFCD00002179
InChI Key VAPDZNUFNKUROY-UHFFFAOYSA-N
Synonym phenol, 2,4,6-triiodo, triiodophenol, unii-9rb2r81a7u, 2,4,6-trijodfenol, 2,4,6-trijodfenol czech, 2,4,6-triiodo-phenol, chembl81721, 2,4,6-triiodo-phenol tip, 2,6-triiodophenol, phenol,4,6-triiodo
PubChem CID 11862
IUPAC Name 2,4,6-triiodophenol
SMILES C1=C(C=C(C(=C1I)O)I)I

Specifications

Melting Point 160°C
Color White
Quantity 5 g
Formula Weight 471.80
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Triiodophenol
RTECSNumber SN2800000
TSCA Yes