missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trihydroxybenzoic Acid Monohydrate 98.0+%, TCI America™
Supplier: TCI America P060125G
Specifications
| 2,4,6-Trihydroxybenzoic Acid Monohydrate | |
| 210°C | |
| MFCD00149095 | |
| 2,4,6-trihydroxybenzoic acid hydrate, 2,4,6-trihydroxybenzoic acid monohydrate, benzoic acid, 2,4,6-trihydroxy-, monohydrate, phloroglucinolcarboxylic acid, pubchem20337, acmc-209olb, 2,4,6-trihydroxybenzoicacidmonohydrate, 2,4,6-trihydroxybenzoic acid, hydrate, 2,4,6-trihydroxybenzoic acid monohydrate, technical t, 2,4,6-trihydroxybenzoic acid monohydrate, technical grade, predominantly 1,3,5-benzenetriol | |
| C1=C(C=C(C(=C1O)C(=O)O)O)O.O | |
| 188.135 | |
| 170.12 | |
| Crystalline Powder |
| 71989-93-0 | |
| C7H8O6 | |
| 25 g | |
| HWZIRFCGHAROOI-UHFFFAOYSA-N | |
| 2,4,6-trihydroxybenzoic acid;hydrate | |
| 2723793 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 71989-93-0 | |
| 188.135 | |
| HWZIRFCGHAROOI-UHFFFAOYSA-N | |
| 2723793 | |
| C1=C(C=C(C(=C1O)C(=O)O)O)O.O |
| C7H8O6 | |
| MFCD00149095 | |
| 2,4,6-trihydroxybenzoic acid hydrate, 2,4,6-trihydroxybenzoic acid monohydrate, benzoic acid, 2,4,6-trihydroxy-, monohydrate, phloroglucinolcarboxylic acid, pubchem20337, acmc-209olb, 2,4,6-trihydroxybenzoicacidmonohydrate, 2,4,6-trihydroxybenzoic acid, hydrate, 2,4,6-trihydroxybenzoic acid monohydrate, technical t, 2,4,6-trihydroxybenzoic acid monohydrate, technical grade, predominantly 1,3,5-benzenetriol | |
| 2,4,6-trihydroxybenzoic acid;hydrate |
Safety and Handling
RTECSNumber : DH8910000
TSCA : Yes