missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trihydroxybenzoic Acid Monohydrate 98.0+%, TCI America™

Catalog No. p-7134693
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06015G 5 g
P060125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P06015G Supplier TCI America Supplier No. P06015G
Only null left

Chemical Identifiers

CAS 71989-93-0
Molecular Formula C7H8O6
Molecular Weight (g/mol) 188.135
MDL Number MFCD00149095
InChI Key HWZIRFCGHAROOI-UHFFFAOYSA-N
Synonym 2,4,6-trihydroxybenzoic acid hydrate, 2,4,6-trihydroxybenzoic acid monohydrate, benzoic acid, 2,4,6-trihydroxy-, monohydrate, phloroglucinolcarboxylic acid, pubchem20337, acmc-209olb, 2,4,6-trihydroxybenzoicacidmonohydrate, 2,4,6-trihydroxybenzoic acid, hydrate, 2,4,6-trihydroxybenzoic acid monohydrate, technical t, 2,4,6-trihydroxybenzoic acid monohydrate, technical grade, predominantly 1,3,5-benzenetriol
PubChem CID 2723793
IUPAC Name 2,4,6-trihydroxybenzoic acid;hydrate
SMILES C1=C(C=C(C(=C1O)C(=O)O)O)O.O

Specifications

Melting Point 210°C
Quantity 5 g
Formula Weight 170.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trihydroxybenzoic Acid Monohydrate
RTECSNumber DH8910000
TSCA Yes