missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',4',6'-Trihydroxyacetophenone Monohydrate 98.0+%, TCI America™
Supplier: TCI America T188825G
Specifications
| 2′,4′,6′-Trihydroxyacetophenone Monohydrate | |
| 215°C | |
| C8H8O4 | |
| 25 g | |
| XLEYFDVVXLMULC-UHFFFAOYSA-N | |
| 1-(2,4,6-trihydroxyphenyl)ethan-1-one | |
| 68073 | |
| 168.15 | |
| Crystalline Powder |
| 480-66-0 | |
| Yellow | |
| MFCD00002287 | |
| 1-2,4,6-trihydroxyphenyl ethanone, 2',4',6'-trihydroxyacetophenone, phloroacetophenone, phloracetophenone, acetophloroglucine, 2,4,6-trihydroxyacetophenone, acetylphloroglucinol, ethanone, 1-2,4,6-trihydroxyphenyl, 2-acetylphloroglucinol, 1-2,4,6-trihydroxyphenyl-ethanone | |
| CC(=O)C1=C(O)C=C(O)C=C1O | |
| 168.15 | |
| CHEBI:64344 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 480-66-0 | |
| 168.15 | |
| XLEYFDVVXLMULC-UHFFFAOYSA-N | |
| 68073 | |
| 1-(2,4,6-trihydroxyphenyl)ethan-1-one |
| C8H8O4 | |
| MFCD00002287 | |
| 1-2,4,6-trihydroxyphenyl ethanone, 2',4',6'-trihydroxyacetophenone, phloroacetophenone, phloracetophenone, acetophloroglucine, 2,4,6-trihydroxyacetophenone, acetylphloroglucinol, ethanone, 1-2,4,6-trihydroxyphenyl, 2-acetylphloroglucinol, 1-2,4,6-trihydroxyphenyl-ethanone | |
| CHEBI:64344 | |
| CC(=O)C1=C(O)C=C(O)C=C1O |
Safety and Handling
RTECSNumber : AN0528000
TSCA : Yes