missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trihydroxyacetophenone monohydrate, Matrix Substance for MALDI-MS, ≥99.5%, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 249278-28-2 |
| Linear Formula | (HO)3C6H2 COCH3·H2O |
| MDL Number | MFCD00149091 |
| Melting Point | 218°C to 222°C; 219°C to 221°C |
| Synonym | 2-Acetylphloroglucinol; THAP |
| Type | Matrix Substance for MALDI-MS |
| Quantity | 5 g |
| Formula Weight | 186.16 |
| Percent Purity | ≥99.5% (HPLC); ≥99.5% |
| Grade | Matrix Substance for MALDI-MS |