missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trihydroxyacetophenone monohydrate, Matrix Substance for MALDI-MS, ≥99.5%, MilliporeSigma™ Supelco™

Catalog No. 111013694
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013694 1 g
111013695 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013694 Supplier Merck Emd Millipore Supplier No. 919281G
Only null left

Specifications

CAS 249278-28-2
Linear Formula (HO)3C6H2 COCH3·H2O
MDL Number MFCD00149091
Melting Point 218°C to 222°C; 219°C to 221°C
Synonym 2-Acetylphloroglucinol; THAP
Type Matrix Substance for MALDI-MS
Quantity 1 g
Formula Weight 186.16
Percent Purity ≥99.5% (HPLC); ≥99.5%
Grade Matrix Substance for MALDI-MS