missing translation for 'onlineSavingsMsg'
Learn More

2',4',6'-Trihydroxyacetophenone hydrate, 98+%

Catalog No. AAL0415606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0415606 5 g
AAL0415614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0415606 Supplier Thermo Scientific Chemicals Supplier No. L0415606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 480-66-0 | C8H8O4 | 168.15 g/mol

2',4',6'-Trihydroxyacetophenone hydrate is a compound that enhances bile secretion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,4′,6′-Trihydroxyacetophenone hydrate is a compound that enhances bile secretion.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 480-66-0
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002287
InChI Key XLEYFDVVXLMULC-UHFFFAOYSA-N
Synonym 1-2,4,6-trihydroxyphenyl ethanone, 2',4',6'-trihydroxyacetophenone, phloroacetophenone, phloracetophenone, acetophloroglucine, 2,4,6-trihydroxyacetophenone, acetylphloroglucinol, ethanone, 1-2,4,6-trihydroxyphenyl, 2-acetylphloroglucinol, 1-2,4,6-trihydroxyphenyl-ethanone
PubChem CID 68073
ChEBI CHEBI:64344
IUPAC Name 1-(2,4,6-trihydroxyphenyl)ethanone
SMILES CC(=O)C1=C(O)C=C(O)C=C1O

Specifications

Melting Point 218°C to 223°C
Quantity 5 g
Beilstein 1911197
Solubility Information Soluble in water.
Formula Weight 168.15 (Anhydrous)
Percent Purity ≥98%
Chemical Name or Material 2',4',6'-Trihydroxyacetophenone hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-556-5
RTECSNumber AN0528000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only