missing translation for 'onlineSavingsMsg'
Learn More

2',4',6'-Trihydroxyacetophenone, 98%

Catalog No. AC139790050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC139790050 5 g
AC139790250 25 g
AC139791000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC139790050 Supplier Thermo Scientific Chemicals Supplier No. 139790050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 480-66-0 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 480-66-0
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002287
InChI Key XLEYFDVVXLMULC-UHFFFAOYSA-N
Synonym 1-2,4,6-trihydroxyphenyl ethanone, 2',4',6'-trihydroxyacetophenone, phloroacetophenone, phloracetophenone, acetophloroglucine, 2,4,6-trihydroxyacetophenone, acetylphloroglucinol, ethanone, 1-2,4,6-trihydroxyphenyl, 2-acetylphloroglucinol, 1-2,4,6-trihydroxyphenyl-ethanone
PubChem CID 68073
ChEBI CHEBI:64344
IUPAC Name 1-(2,4,6-trihydroxyphenyl)ethanone
SMILES CC(=O)C1=C(O)C=C(O)C=C1O

Specifications

Melting Point 219.0°C to 222.0°C
Color Brown to Orange-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (HO)3C6H2COCH3
Quantity 5 g
Beilstein 08,II,442
Formula Weight 168.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2', 4', 6'-Trihydroxyacetophenone
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 207-556-5

RUO – Research Use Only