missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trifluoronitrobenzene 97.0+%, TCI America™
Supplier: TCI America T16815G
Specifications
| 2,4,6-Trifluoronitrobenzene | |
| Yellow | |
| C6H2F3NO2 | |
| 5 g | |
| PWRFDGYYJWQIAB-UHFFFAOYSA-N | |
| 1,3,5-trifluoro-2-nitrobenzene | |
| 67567 | |
| ≥97.0% (GC) |
| 315-14-0 | |
| 172°C | |
| MFCD00014687 | |
| 2,4,6-trifluoronitrobenzene, 2,2,6-trifluoronitrobenzene, benzene, 1,3,5-trifluoro-2-nitro, 1,3,5-trifluoro-2-nitro-benzene, 2-nitro-1,3,5-trifluorobenzene, pubchem10646, acmc-209hn4, 1,3,5-trifluoronitrobenzene, 2,4,6 trifluoronitrobenzene, ksc226o4t | |
| C1=C(C=C(C(=C1F)[N+](=O)[O-])F)F | |
| 177.082 | |
| 177.08 | |
| Liquid |
Chemical Identifiers
| 315-14-0 | |
| 177.082 | |
| PWRFDGYYJWQIAB-UHFFFAOYSA-N | |
| 67567 | |
| C1=C(C=C(C(=C1F)[N+](=O)[O-])F)F |
| C6H2F3NO2 | |
| MFCD00014687 | |
| 2,4,6-trifluoronitrobenzene, 2,2,6-trifluoronitrobenzene, benzene, 1,3,5-trifluoro-2-nitro, 1,3,5-trifluoro-2-nitro-benzene, 2-nitro-1,3,5-trifluorobenzene, pubchem10646, acmc-209hn4, 1,3,5-trifluoronitrobenzene, 2,4,6 trifluoronitrobenzene, ksc226o4t | |
| 1,3,5-trifluoro-2-nitrobenzene |
Safety and Handling
TSCA : No