missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorobenzoic Acid 98.0+%, TCI America™

Catalog No. T24091G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24091G Supplier TCI America Supplier No. T24091G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 28314-80-9
Molecular Formula C7H3F3O2
Molecular Weight (g/mol) 176.094
MDL Number MFCD00042398
InChI Key SJZATRRXUILGHH-UHFFFAOYSA-N
Synonym benzoic acid, 2,4,6-trifluoro, 2,4,6-trifluorobenzoicacid, the derivatives, 2,4,6-trifluorobenzioc acid, pubchem1347, acmc-209h1r, ksc204g9t, 2,4,6-trifluoro-benzoic acid, rarechem al bo 0448, benzoicacid, 2,4,6-trifluoro
PubChem CID 520373
IUPAC Name 2,4,6-trifluorobenzoic acid
SMILES C1=C(C=C(C(=C1F)C(=O)O)F)F

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 1 g
Formula Weight 176.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trifluorobenzoic Acid