missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorobenzoic acid, 98%

Catalog No. p-7024673
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAA1975803 1 g
AAA1975806 5 g
2 options

Catalog No. AAA1975803

Supplier: Thermo Scientific Chemicals A1975803

Only null left

CAS: 28314-80-9 | C7H3F3O2 | 176.094 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28314-80-9
Molecular Formula C7H3F3O2
Molecular Weight (g/mol) 176.094
MDL Number MFCD00042398
InChI Key SJZATRRXUILGHH-UHFFFAOYSA-N
Synonym benzoic acid, 2,4,6-trifluoro, 2,4,6-trifluorobenzoicacid, the derivatives, 2,4,6-trifluorobenzioc acid, pubchem1347, acmc-209h1r, ksc204g9t, 2,4,6-trifluoro-benzoic acid, rarechem al bo 0448, benzoicacid, 2,4,6-trifluoro
PubChem CID 520373
IUPAC Name 2,4,6-trifluorobenzoic acid
SMILES C1=C(C=C(C(=C1F)C(=O)O)F)F

Specifications

Melting Point 140°C to 142°C
Quantity 1 g
Beilstein 1958300
Formula Weight 176.1
Percent Purity 98%
Chemical Name or Material 2,4,6-Trifluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only