missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorobenzenesulfonyl chloride, 97%

CAS: 220239-64-5 | C6H2ClF3O2S | 230.585 g/mol

Supplier:  Thermo Scientific Chemicals H3274603

 View more versions of this product

Catalog No. AAH3274603


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4,6-Trifluorobenzenesulfonyl chloride
C6H2ClF3O2S
1 g
Moisture sensitive
XINCBNCLCIQIJM-UHFFFAOYSA-N
2,4,6-trifluorobenzenesulfonyl chloride
2776983
97%
220239-64-5
MFCD01091015
UN3265
2,4,6-trifluorobenzene-1-sulfonyl chloride, 2,4,6-trifluorobenzenesulphonyl chloride, pubchem5181, chloro 2,4,6-trifluorophenyl sulfone, 2,4,6-trifluorobenzenesulfonylchloride, 2,4,6-trifluorophenylsulfonyl chloride, benzenesulfonylchloride, 2,4,6-trifluoro, 2,4,6-trifluorobenzenesulfonyl chloride, benzenesulfonyl chloride, 2,4,6-trifluoro-9ci
C1=C(C=C(C(=C1F)S(=O)(=O)Cl)F)F
230.585
230.59

Chemical Identifiers

220239-64-5
230.585
XINCBNCLCIQIJM-UHFFFAOYSA-N
2776983
C1=C(C=C(C(=C1F)S(=O)(=O)Cl)F)F
C6H2ClF3O2S
MFCD01091015
2,4,6-trifluorobenzene-1-sulfonyl chloride, 2,4,6-trifluorobenzenesulphonyl chloride, pubchem5181, chloro 2,4,6-trifluorophenyl sulfone, 2,4,6-trifluorobenzenesulfonylchloride, 2,4,6-trifluorophenylsulfonyl chloride, benzenesulfonylchloride, 2,4,6-trifluoro, 2,4,6-trifluorobenzenesulfonyl chloride, benzenesulfonyl chloride, 2,4,6-trifluoro-9ci
2,4,6-trifluorobenzenesulfonyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only