missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorobenzenesulfonyl chloride, 97%

Catalog No. p-7031612
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32746MD 250 mg
AAH3274603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32746MD Supplier Thermo Scientific Chemicals Supplier No. H32746MD
Only null left

CAS: 220239-64-5 | C6H2ClF3O2S | 230.585 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 220239-64-5
Molecular Formula C6H2ClF3O2S
Molecular Weight (g/mol) 230.585
MDL Number MFCD01091015
InChI Key XINCBNCLCIQIJM-UHFFFAOYSA-N
Synonym 2,4,6-trifluorobenzene-1-sulfonyl chloride, 2,4,6-trifluorobenzenesulphonyl chloride, pubchem5181, chloro 2,4,6-trifluorophenyl sulfone, 2,4,6-trifluorobenzenesulfonylchloride, 2,4,6-trifluorophenylsulfonyl chloride, benzenesulfonylchloride, 2,4,6-trifluoro, 2,4,6-trifluorobenzenesulfonyl chloride, benzenesulfonyl chloride, 2,4,6-trifluoro-9ci
PubChem CID 2776983
IUPAC Name 2,4,6-trifluorobenzenesulfonyl chloride
SMILES C1=C(C=C(C(=C1F)S(=O)(=O)Cl)F)F

Specifications

Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 230.59
Percent Purity 97%
Chemical Name or Material 2,4,6-Trifluorobenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only