missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorobenzaldehyde 98.0+%, TCI America™

Catalog No. T23201G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23201G 1 g
T23205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23201G Supplier TCI America Supplier No. T23201G
Only null left

Chemical Identifiers

CAS 58551-83-0
Molecular Formula C7H3F3O
Molecular Weight (g/mol) 160.095
MDL Number MFCD00061197
InChI Key KPJIEPBITZLHPQ-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4,6-trifluoro, 2,4,6-ttrifluorobenzaldehyde, pubchem1416, acmc-209m5d, 2,4,6 trifluorobenzaldehyde, ksc272c8l, 2,4,6-trifluoro benzaldehyde, 2,4,6-trifluoro-benzaldehyde
PubChem CID 521845
IUPAC Name 2,4,6-trifluorobenzaldehyde
SMILES C1=C(C=C(C(=C1F)C=O)F)F

Specifications

Melting Point 66°C
Quantity 1 g
Formula Weight 160.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trifluorobenzaldehyde