missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichlorophenyl Formate 97.0+%, TCI America™

Catalog No. T31211G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31211G 1 g
T31215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31211G Supplier TCI America Supplier No. T31211G

Chemical Identifiers

CAS 4525-65-9
Molecular Formula C7H3Cl3O2
Molecular Weight (g/mol) 225.449
InChI Key YSUDXADDVAYVNS-UHFFFAOYSA-N
Synonym Formic Acid 2,4,6-Trichlorophenyl Ester
PubChem CID 53750963
IUPAC Name (2,4,6-trichlorophenyl) formate
SMILES C1=C(C=C(C(=C1Cl)OC=O)Cl)Cl

Specifications

Melting Point 83°C
Color White
Quantity 1 g
Formula Weight 225.45
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trichlorophenyl Formate