missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichlorobenzoyl Chloride 98.0+%, TCI America™

Catalog No. p-7136764
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
T14135G 5 g
T141325G 25 g
2 options

Catalog No. T14135G

Supplier: TCI America T14135G

Only null left

Chemical Identifiers

CAS 4136-95-2
Molecular Formula C7H2Cl4O
Molecular Weight (g/mol) 243.89
MDL Number MFCD00075323
InChI Key OZGSEIVTQLXWRO-UHFFFAOYSA-N
Synonym 2,4,6-trichlorobenzoylchloride, benzoyl chloride, 2,4,6-trichloro, mi, acmc-209jk3, ksc494s2p, 2,4,6-cl3c6h2cocl, 2,4,6-ttrichlorobenzoyl chloride, 2,4,6-trichloro benzoyl chloride, 2,4,6-trichlorobenzoic acid chloride, 2,4,6-tris chloranyl benzoyl chloride
PubChem CID 2733703
IUPAC Name 2,4,6-trichlorobenzoyl chloride
SMILES ClC(=O)C1=C(Cl)C=C(Cl)C=C1Cl

Specifications

Color Yellow
Boiling Point 275°C
Quantity 5 g
UN Number 3265
Formula Weight 243.89
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,4,6-Trichlorobenzoyl Chloride
EINECSNumber (3)-0062
TSCA No