missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichlorobenzoyl chloride, 98%

Catalog No. p-7058411
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1415906 5 g
AAL1415914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1415906 Supplier Thermo Scientific Chemicals Supplier No. L1415906
Only null left

CAS: 4136-95-2 | C7H2Cl4O | 243.89 g/mol

2,4,6-Trichlorobenzoyl chloride is used in the preparation of γ-lactone and δ-lactone, aliphatic aromatic anhydrides, required for the synthesis of amphiphilic hyaluronan, mixed anhydride, required for the synthesis of angelate esters, synthesis of both spongistatin 1 and spongistatin 2 and large-ring lactones in high yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trichlorobenzoyl chloride is used in the preparation of γ-lactone and δ-lactone, aliphatic aromatic anhydrides, required for the synthesis of amphiphilic hyaluronan, mixed anhydride, required for the synthesis of angelate esters, synthesis of both spongistatin 1 and spongistatin 2 and large-ring lactones in high yields.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to moisture. Incompatible with strong bases and strong oxidizing agents.

Chemical Identifiers

CAS 4136-95-2
Molecular Formula C7H2Cl4O
Molecular Weight (g/mol) 243.89
MDL Number MFCD00075323
InChI Key OZGSEIVTQLXWRO-UHFFFAOYSA-N
Synonym 2,4,6-trichlorobenzoylchloride, benzoyl chloride, 2,4,6-trichloro, mi, acmc-209jk3, ksc494s2p, 2,4,6-cl3c6h2cocl, 2,4,6-ttrichlorobenzoyl chloride, 2,4,6-trichloro benzoyl chloride, 2,4,6-trichlorobenzoic acid chloride, 2,4,6-tris chloranyl benzoyl chloride
PubChem CID 2733703
IUPAC Name 2,4,6-trichlorobenzoyl chloride
SMILES ClC(=O)C1=C(Cl)C=C(Cl)C=C1Cl

Specifications

Density 1.561
Boiling Point 107°C to 108°C (6 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.575
Quantity 5 g
UN Number UN3265
Beilstein 2050280
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 243.91
Percent Purity 98%
Chemical Name or Material 2,4,6-Trichlorobenzoyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only