missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trichloroanisole 98.0+%, TCI America™
Supplier: TCI America T086725G
Specifications
| 2,4,6-Trichloroanisole | |
| 60°C | |
| 240°C | |
| MFCD00000588 | |
| 2,4,6-trichloroanisole, tyrene, trichloroanisole, 2,4,6-trichloro-1-methoxybenzene, benzene, 1,3,5-trichloro-2-methoxy, methyl 2,4,6-trichlorophenyl ether, 2,4.6-trichloroanisole, anisole, 2,4,6-trichloro, anisole, 2,4,6-trichloro-8ci, benzene,1,3,5-trichloro-2-methoxy | |
| COC1=C(C=C(C=C1Cl)Cl)Cl | |
| 211.466 | |
| CHEBI:19333 | |
| ≥98.0% (GC) |
| 87-40-1 | |
| White | |
| C7H5Cl3O | |
| 25 g | |
| WCVOGSZTONGSQY-UHFFFAOYSA-N | |
| 1,3,5-trichloro-2-methoxybenzene | |
| 6884 | |
| 211.47 | |
| Crystalline Powder |
Chemical Identifiers
| 87-40-1 | |
| 211.466 | |
| WCVOGSZTONGSQY-UHFFFAOYSA-N | |
| CHEBI:19333 | |
| COC1=C(C=C(C=C1Cl)Cl)Cl |
| C7H5Cl3O | |
| MFCD00000588 | |
| 6884 | |
| 1,3,5-trichloro-2-methoxybenzene |
Safety and Handling
TSCA : No