missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trichloroanisole Solution, Certified Reference Material, 100 μg/mL in Methanol, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Ampule |
| CAS | 87-40-1 |
| Product Line | TraceCERT |
| Concentration | 100 μg/mL in methanol |
| Type | Food and Cosmetic Component Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC