missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichloro-5-methylpyrimidine, 98%

Catalog No. p-7036756
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5511503 1 g
AAH5511506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5511503 Supplier Thermo Scientific Chemicals Supplier No. H5511503
Only null left

CAS: 1780-36-5 | C5H3Cl3N2 | 197.443 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1780-36-5
Molecular Formula C5H3Cl3N2
Molecular Weight (g/mol) 197.443
MDL Number MFCD00233525
InChI Key VTSWSQGDJQFXHB-UHFFFAOYSA-N
Synonym 5-methyl-2,4,6-trichloropyrimidine, pyrimidine, 2,4,6-trichloro-5-methyl, 2,4,6-trichlor-5-methylpyrimidin, pubchem7341, acmc-20a0tr, 2,4,6-trichloro-5, ksc179k8l, 2,6-trichloro-5-methylpyrimidine, methyl 2,4,6-trichloropyrimidine, 2,4,6-trichoro-5-methylpyrimidine
PubChem CID 232789
IUPAC Name 2,4,6-trichloro-5-methylpyrimidine
SMILES CC1=C(N=C(N=C1Cl)Cl)Cl

Specifications

Melting Point 65°C to 69°C
Quantity 1 g
UN Number UN1759
Formula Weight 197.45
Percent Purity 98%
Chemical Name or Material 2,4,6-Trichloro-5-methylpyrimidine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only