missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromotoluene, 98+%

Catalog No. AAL0114909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0114909 10 g
AAL0114918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0114909 Supplier Thermo Scientific Chemicals Supplier No. L0114909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6320-40-7 | C7H5Br3 | 328.829 g/mol

2,4,6-Tribromotoluene, is used as a chemical and organic intermediate. It can be used to produce 2,4,6-tribromo-benzyl bromide at temperature of 200 - 220°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Tribromotoluene, is used as a chemical and organic intermediate. It can be used to produce 2,4,6-tribromo-benzyl bromide at temperature of 200 - 220°C.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 6320-40-7
Molecular Formula C7H5Br3
Molecular Weight (g/mol) 328.829
MDL Number MFCD00013527
InChI Key BFRIZWKDNUHPHL-UHFFFAOYSA-N
Synonym 2,4,6-tribromotoluene, benzene, 1,3,5-tribromo-2-methyl, acmc-20aof7, 2,4,6,tribromotoluene, 2,4,6-tribrom-toluol, 2-methyl-1,3,5-tribromobenzene, benzene,1,3,5-tribromo-2-methyl, 2,4,6-tribromotoluene 10g, 1,3,5-tribromo-2-methyl-benzene
PubChem CID 33916
IUPAC Name 1,3,5-tribromo-2-methylbenzene
SMILES CC1=C(C=C(C=C1Br)Br)Br

Specifications

Melting Point 67°C to 70°C
Density 2.479
Boiling Point 290°C
Quantity 10 g
Beilstein 2359313
Solubility Information Insoluble in water.
Formula Weight 328.83
Percent Purity ≥98%
Chemical Name or Material 2,4,6-Tribromotoluene
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 228-672-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only