missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromoresorcinol 98.0+%, TCI America™

Catalog No. p-7137322
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26201G 1 g
T26205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26201G Supplier TCI America Supplier No. T26201G
Only null left

Chemical Identifiers

CAS 2437-49-2
Molecular Formula C6H3Br3O2
Molecular Weight (g/mol) 346.8
MDL Number MFCD00009717
InChI Key YADZSMVDNYXOOB-UHFFFAOYSA-N
Synonym 2,4,6-tribromoresorcinol, resorcinol, 2,4,6-tribromo, 1,3-benzenediol, 2,4,6-tribromo, 2,6-tribromoresorcinol, acmc-209gbn, 2,4,6-tribromresorcin, 2,4,6-tribromoresorcino, resorcinol,4,6-tribromo, 1, 2,4,6-tribromo, 4-06-00-05688 beilstein handbook reference
PubChem CID 17094
IUPAC Name 2,4,6-tribromobenzene-1,3-diol
SMILES C1=C(C(=C(C(=C1Br)O)Br)O)Br

Specifications

Melting Point 114°C
Quantity 1 g
Formula Weight 346.80
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tribromoresorcinol
RTECSNumber VH3500000
TSCA No