missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromoresorcinol, 98%

Catalog No. AAA1943603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1943603 1 g
AAA1943606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1943603 Supplier Thermo Scientific Chemicals Supplier No. A1943603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2437-49-2 | C6H3Br3O2 | 346.8 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2437-49-2
Molecular Formula C6H3Br3O2
Molecular Weight (g/mol) 346.8
MDL Number MFCD00009717
InChI Key YADZSMVDNYXOOB-UHFFFAOYSA-N
Synonym 2,4,6-tribromoresorcinol, resorcinol, 2,4,6-tribromo, 1,3-benzenediol, 2,4,6-tribromo, 2,6-tribromoresorcinol, acmc-209gbn, 2,4,6-tribromresorcin, 2,4,6-tribromoresorcino, resorcinol,4,6-tribromo, 1, 2,4,6-tribromo, 4-06-00-05688 beilstein handbook reference
PubChem CID 17094
IUPAC Name 2,4,6-tribromobenzene-1,3-diol
SMILES C1=C(C(=C(C(=C1Br)O)Br)O)Br

Specifications

Melting Point 114°C to 116°C
Quantity 1 g
Formula Weight 346.82
Percent Purity 98%
Chemical Name or Material 2,4,6-Tribromoresorcinol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-442-2
RTECSNumber VH3500000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only