missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromophenol, 98%

CAS: 118-79-6 | C6H3Br3O | 330.80 g/mol

Supplier:  Thermo Scientific Chemicals 160101000

 View more versions of this product

Catalog No. AC160101000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 4, 6-Tribromophenol
118-79-6
100.0
White to Brown
Authentic
Plastic bottle
Br3C6H2OH
100 g
02,424
tribromophenol, bromol, bromkal pur 3, xeroform, phenol, 2,4,6-tribromo, flammex 3bp, unii-ys6k3eu393, ccris 1658, great lakes ph-73, 5175-83-7 bismuth 3+ salt
Solubility in water: 71mg/l (15°C). Other solubilities: soluble in acetone,diethylether,petroleum ether,,benzene,toluene,alcohols,chloroform,pyridine,and carbon tetrachloride
OC1=C(Br)C=C(Br)C=C1Br
330.80
CHEBI:47696
98%
98%
97.5
89.0°C to 93.0°C
244.0°C
98%
C6H3Br3O
MFCD00002150
06,203
15,9775
244°C
BSWWXRFVMJHFBN-UHFFFAOYSA-N
2,4,6-tribromophenol
1483
330.79
Powder

Chemical Identifiers

118-79-6
330.80
BSWWXRFVMJHFBN-UHFFFAOYSA-N
1483
2,4,6-tribromophenol
C6H3Br3O
MFCD00002150
tribromophenol, bromol, bromkal pur 3, xeroform, phenol, 2,4,6-tribromo, flammex 3bp, unii-ys6k3eu393, ccris 1658, great lakes ph-73, 5175-83-7 bismuth 3+ salt
CHEBI:47696
OC1=C(Br)C=C(Br)C=C1Br
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Causes serious eye irritation.
Very toxic to aquatic life.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/ey

GHS Signal Word: Warning

EINECSNumber : 204-278-6

RTECSNumber : SN1225000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only