missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromophenol, 98%

Catalog No. AC160100050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160100050 5 g Glass Bottle
AC160101000 100 g Plastic bottle
AC160105000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC160100050 Supplier Thermo Scientific Chemicals Supplier No. 160100050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 118-79-6 | C6H3Br3O | 330.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-79-6
Molecular Formula C6H3Br3O
Molecular Weight (g/mol) 330.80
MDL Number MFCD00002150
InChI Key BSWWXRFVMJHFBN-UHFFFAOYSA-N
Synonym tribromophenol, bromol, bromkal pur 3, xeroform, phenol, 2,4,6-tribromo, flammex 3bp, unii-ys6k3eu393, ccris 1658, great lakes ph-73, 5175-83-7 bismuth 3+ salt
PubChem CID 1483
ChEBI CHEBI:47696
IUPAC Name 2,4,6-tribromophenol
SMILES OC1=C(Br)C=C(Br)C=C1Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 89.0°C to 93.0°C
Color White to Brown
Boiling Point 244.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula Br3C6H2OH
Quantity 5 g
Beilstein 06,203
Fieser 02,424
Merck Index 15,9775
Solubility Information Solubility in water: 71mg/l (15°C). Other solubilities: soluble in acetone,diethylether,petroleum ether,,benzene,toluene,alcohols,chloroform,pyridine,and carbon tetrachloride
Sublimation Point 244°C
Formula Weight 330.79
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2, 4, 6-Tribromophenol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Causes serious eye irritation.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/ey
EINECSNumber 204-278-6
RTECSNumber SN1225000
TSCA TSCA

RUO – Research Use Only