missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Tribromobenzoic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 192°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 358.81 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,4,6-Tribromobenzoic Acid |
Safety and Handling
| TSCA | No |