missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromobenzoic Acid 98.0+%, TCI America™

Catalog No. T06191G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T06191G 1 g
T06195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T06191G Supplier TCI America Supplier No. T06191G

Chemical Identifiers

CAS 633-12-5
Molecular Formula C7H3Br3O2
Molecular Weight (g/mol) 358.811
MDL Number MFCD00059882
InChI Key GHVMJSHEGZYRQL-UHFFFAOYSA-N
PubChem CID 246577
IUPAC Name 2,4,6-tribromobenzoic acid
SMILES C1=C(C=C(C(=C1Br)C(=O)O)Br)Br

Specifications

Melting Point 192°C
Color White-Yellow
Quantity 1 g
Formula Weight 358.81
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tribromobenzoic Acid