missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromo-3-hydroxybenzoic acid, 97%

Catalog No. p-7027212
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2379006 5 g
AAB2379014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2379006 Supplier Thermo Scientific Chemicals Supplier No. B2379006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14348-40-4 | C7H3Br3O3 | 374.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14348-40-4
Molecular Formula C7H3Br3O3
Molecular Weight (g/mol) 374.81
MDL Number MFCD00055557
InChI Key YDBHVMTTYXWHLI-UHFFFAOYSA-N
Synonym 3-hydroxy-2,4,6-tribromobenzoic acid, tbhba, benzoic acid, 2,4,6-tribromo-3-hydroxy, 2,4,6-tribromo-3-hydroxybenzoicacid, 2,4,6-thba, pubchem12541, acmc-209uep, ksc174o3h, 2,4,6-tribromo-3-hydroxy-benzoic acid, 2,4,6-tribromo-3-hydroxybenzoic acid
PubChem CID 151915
IUPAC Name 2,4,6-tribromo-3-hydroxybenzoic acid
SMILES C1=C(C(=C(C(=C1Br)O)Br)C(=O)O)Br

Specifications

Melting Point 144°C to 149°C
Quantity 5 g
Beilstein 2844497
Formula Weight 374.81
Percent Purity 98%
Chemical Name or Material 2,4,6-Tribromo-3-hydroxybenzoic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only