missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triaminopyrimidine 98.0+%, TCI America™

Catalog No. p-7136451
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T083425G 25 g
T0834500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T083425G Supplier TCI America Supplier No. T083425G
Only null left

Chemical Identifiers

CAS 1004-38-2
Molecular Formula C4H7N5
Molecular Weight (g/mol) 125.135
MDL Number MFCD00006100
InChI Key JTTIOYHBNXDJOD-UHFFFAOYSA-N
Synonym 2,4,6-triaminopyrimidine, 2,4,6-pyrimidinetriamine, pyrimidine-2,4,6-triyltriamine, pyrimidine, 2,4,6-triamino, triamino pyrimidine, 3ay, aaat, pubchem7216, 2,6-pyrimidinetriamine, 2,6-triaminopyrimidine
PubChem CID 13863
ChEBI CHEBI:39857
IUPAC Name pyrimidine-2,4,6-triamine
SMILES C1=C(N=C(N=C1N)N)N

Specifications

Melting Point 249°C
Color Yellow
Quantity 25 g
Formula Weight 125.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Triaminopyrimidine