accessibility menu, dialog, popup

UserName

NH2NH2NH2NN

CAS RN 1004-38-2

IUPAC Name: pyrimidine-2,4,6-triamine
Synonyms: 2,4,6-triaminopyrimidine
Molecular Weight (g/mol): 125.14
Molecular Formula: C4H7N5
InChi Key: JTTIOYHBNXDJOD-UHFFFAOYSA-N
SMILES: NC1=CC(N)=NC(N)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results