missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Triaminopyrimidine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 249°C |
| Color | Yellow |
| Quantity | 500 g |
| Formula Weight | 125.14 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,4,6-Triaminopyrimidine |
Safety and Handling
| TSCA | Yes |