missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triaminopyrimidine, 97%

Catalog No. p-202224
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC139250250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC139250250 Supplier Thermo Scientific Chemicals Supplier No. 139250250
Only null left

CAS: 1004-38-2 | C4H7N5 | 125.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1004-38-2
MDL Number MFCD00006100
InChI Key JTTIOYHBNXDJOD-UHFFFAOYSA-N
Synonym 2,4,6-triaminopyrimidine, 2,4,6-pyrimidinetriamine, pyrimidine-2,4,6-triyltriamine, pyrimidine, 2,4,6-triamino, triamino pyrimidine, 3ay, aaat, pubchem7216, 2,6-pyrimidinetriamine, 2,6-triaminopyrimidine
PubChem CID 13863
ChEBI CHEBI:39857
IUPAC Name pyrimidine-2,4,6-triamine
SMILES C1=C(N=C(N=C1N)N)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 246°C to 252°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Molecular Formula C4H7N5
Quantity 25 g
Beilstein 24,47
Solubility Information Solubility in water: 36.5g/kg in water (20°C). Other solubilities: 22.7g/kg in methanol (20°C),4.5g/kg in ethanol (20°C)
Molecular Weight (g/mol) 125.13
Formula Weight 125.13
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 4, 6-Triaminopyrimidine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-720-7
TSCA TSCA

RUO – Research Use Only