missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tri-tert-butylnitrosobenzene 98.0+%, TCI America™

Catalog No. T1534100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1534100MG 100 mg
T15341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1534100MG Supplier TCI America Supplier No. T1534100MG

Chemical Identifiers

CAS 24973-59-9
Molecular Formula C18H29NO
Molecular Weight (g/mol) 275.44
MDL Number MFCD00008817
InChI Key OSICDPWAPKXXHT-UHFFFAOYSA-N
PubChem CID 90676
IUPAC Name 1,3,5-tri-tert-butyl-2-nitrosobenzene
SMILES CC(C)(C)C1=CC(=C(N=O)C(=C1)C(C)(C)C)C(C)(C)C

Specifications

Melting Point 168°C
Color Green
Quantity 100 mg
Synonym 2,4,6-tri-tert-butylnitrosobenzene, 1,3,5-tri-tert-butyl-2-nitrosobenzene, 2,4,6-tri-t-butyl-nitrosobenzene, ttbnb, 1,3,5-tri tert-butyl-2-nitrosobenzene, benzene, 1,3,5-tris 1,1-dimethylethyl-2-nitroso, 2-nitroso-1,3,5-tri-tert-butylbenzene, benzene,1,3,5-tris 1,1-dimethylethyl-2-nitroso, acmc-1cr76, 2,4,6-tri-t-butylnitrosobenzene
Formula Weight 275.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tri-tert-butylnitrosobenzene
TSCA Yes
Recommended Storage Refrigerator