missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tri-tert-butylaniline 98.0+%, TCI America™

Catalog No. p-7136915
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16941G 1 g
T16945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16941G Supplier TCI America Supplier No. T16941G
Only null left

Chemical Identifiers

CAS 961-38-6
Molecular Formula C18H31N
Molecular Weight (g/mol) 261.453
MDL Number MFCD00011645
InChI Key REJGDSCBQPJPQT-UHFFFAOYSA-N
Synonym 2,4,6-tri-tert-butylaniline, 2,4,6-tri-t-butylaniline, 2,4,6-tri tert-butyl aniline, aniline, tri-tert-butyl, 2,4,6-tri-tert-butylphenylamine, 2,4,6-tritert-butylphenylamine, benzenamine, 2,4,6-tris 1,1-dimethylethyl, aniline, 2,4,6-tri-tert-butyl, 2,4,6-tris tert-butyl phenylamine, pubchem16758
PubChem CID 70402
IUPAC Name 2,4,6-tritert-butylaniline
SMILES CC(C)(C)C1=CC(=C(C(=C1)C(C)(C)C)N)C(C)(C)C

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 1 g
Formula Weight 261.45
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tri-tert-butylaniline