missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tri(9H-carbazol-9-yl)-1,3,5-triazine (purified by sublimation) 98.0+%, TCI America™

Catalog No. T27001G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27001G 1 g
T27005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27001G Supplier TCI America Supplier No. T27001G
Only null left

Chemical Identifiers

CAS 134984-37-5
Molecular Formula C39H24N6
Molecular Weight (g/mol) 576.66
MDL Number MFCD18207722
InChI Key NPLMKKHOVKWGEO-UHFFFAOYSA-N
PubChem CID 23399531
IUPAC Name 9-[4,6-bis(9H-carbazol-9-yl)-1,3,5-triazin-2-yl]-9H-carbazole
SMILES C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=NC(=NC(=N1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Color White
Quantity 1 g
Formula Weight 576.66
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tri(9H-carbazol-9-yl)-1,3,5-triazine (purified by sublimation)