CAS RN 134984-37-5
CAS RN 134984-37-5
IUPAC Name:
9-[4,6-bis(9H-carbazol-9-yl)-1,3,5-triazin-2-yl]-9H-carbazole
Synonyms:
9-[4,6-bis(carbazol-9-yl)-1,3,5-triazin-2-yl]carbazole
Molecular Weight (g/mol):
576.66
Molecular Formula:
C39H24N6
InChi Key:
NPLMKKHOVKWGEO-UHFFFAOYSA-N
SMILES:
C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=NC(=NC(=N1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)N1C2=C(C=CC=C2)C2=C1C=CC=C2
1
–
1
of
1
results
1
2,4,6-Tri(9H-carbazol-9-yl)-1,3,5-triazine (purified by sublimation) 98.0+%, TCI America™
CAS: 134984-37-5 Molecular Formula: C39H24N6 Molecular Weight (g/mol): 576.66 MDL Number: MFCD18207722 InChI Key: NPLMKKHOVKWGEO-UHFFFAOYSA-N PubChem CID: 23399531 IUPAC Name: 9-[4,6-bis(9H-carbazol-9-yl)-1,3,5-triazin-2-yl]-9H-carbazole SMILES: C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=NC(=NC(=N1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)N1C2=C(C=CC=C2)C2=C1C=CC=C2