missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tri([1,1'-biphenyl]-4-yl)-1,3,5-triazine 98.0+%, TCI America™

Catalog No. T3539200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3539200MG 200 mg
T35391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3539200MG Supplier TCI America Supplier No. T3539200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31274-51-8
Molecular Formula C39H27N3
Molecular Weight (g/mol) 537.666
InChI Key CENPSTJGQOQKKW-UHFFFAOYSA-N
PubChem CID 11628027
IUPAC Name 2,4,6-tris(4-phenylphenyl)-1,3,5-triazine
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=NC(=NC(=N3)C4=CC=C(C=C4)C5=CC=CC=C5)C6=CC=C(C=C6)C7=CC=CC=C7

Specifications

Melting Point 282°C
Color Yellow
Quantity 200 mg
Formula Weight 537.67
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tri([1,1′-biphenyl]-4-yl)-1,3,5-triazine